Skip to Content
Merck
All Photos(1)

Key Documents

AX0110

Supelco

Acetone

OmniSolv® HR-GC

Synonym(s):

Dimethyl Ketone, 2-Propanone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH3
CAS Number:
Molecular Weight:
58.08
Beilstein:
635680
MDL number:
UNSPSC Code:
12352115
EC Index Number:
200-662-2
Assay:
≥99.7% (GC)
technique(s):
HPLC: suitable
LC/MS: suitable
gas chromatography (GC): suitable
bp:
56 °C/760 mmHg (lit.)
vapor pressure:
184 mmHg ( 20 °C)

Agency

suitable for EPA 1633

Quality Level

vapor density

2 (vs air)

vapor pressure

184 mmHg ( 20 °C)

product line

OmniSolv®

Assay

≥99.7% (GC)

form

liquid

expl. lim.

13.2 %

technique(s)

HPLC: suitable
LC/MS: suitable
gas chromatography (GC): suitable

impurities

≤0.3 μeq/g Titrable acid
≤0.5% Water (H2O)

evapn. residue

≤1 ppm

color

APHA: ≤5

refractive index

n20/D 1.359 (lit.)

pH

5-6 (20 °C, 395 g/L in H2O)

bp

56 °C/760 mmHg (lit.)

mp

−94 °C (lit.)

transition temp

flash point <-20 °C

density

0.791 g/mL at 25 °C (lit.)

format

neat

shipped in

ambient

storage temp.

room temp

SMILES string

CC(C)=O

InChI

1S/C3H6O/c1-3(2)4/h1-2H3

InChI key

CSCPPACGZOOCGX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Acetone′s luminesence intensity is dependent upon the solution components . The absorption of UV light by acetone, results in its photolysis and the production of radials .
OmniSolv® HR-GC

Preparation Note

Product filtered through a 0.2 μm filter

Analysis Note

AppearanceClear liquid, free from particulatesAssay (GC)99.7% minCap. ECD responsive substances as PCNB (peak)2ng/L maxCap. ECD responsive substances as PCNB (total)50ng/L maxCap. FID responsive substances as decane (peak)1µg/L maxCap. FID responsive substances as decane (total)10µg/L maxColor (APHA)5 maxEvaporation residue1ppm maxFiltered through 0.2 µm filterTo pass testTitrable acid0.3µeq/g maxWater (H2O)0.5% max

Legal Information

OMNISOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service