Skip to Content
Merck
All Photos(1)

Key Documents

ABN910

Sigma-Aldrich

Anti-Aquaporin 4

from rabbit, purified by affinity chromatography

Synonym(s):

AQP-4, Mercurial-insensitive water channel, MIWC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse, rat

packaging

antibody small pack of 25 μg

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... AQP4(361)
mouse ... Aqp4(11829)
rat ... Aqp4(25293)

General description

Aquaporin-4 (UniProt: P47863; also known as AQP-4, Mercurial-insensitive water channel, MIWC) is encoded by the Aqp4 gene (Gene ID: 25293) in rat. Aquaporin-4 is a multi-pass membrane homotetrameric protein that forms a water-specific channel and serves as an osmoreceptor that regulates body water balance and mediates water flow within the central nervous system. Its extracellular domains are localized to amino acids 58-64, 137-155, and 206-231); its transmembrane domains are in amino acids 37-57, 65-85, 116-136, 156-176, 185-205, and 232-252); and the cytoplasmic domain is in amino acids 1-36, 86-115, 177-184, and 253-323). Aquaporin-4 is largely expressed in the brain and the skeletal muscle and at lower levels in heart, kidney, and lung tissue. Phosphorylation of Aquaporin-4 at Serine 180 by protein kinase C is shown to reduce conductance by about 50%, whereas phosphorylation at Serine 111 by protein kinase G in response to glutamate can increase conductance by about 40%. Two isoforms of Aquaporin-4 have been described that are produced by alternative splicing. Antibodies to Aquaporin-4 can serve as useful serum markers to distinguishing neuromyelitis optica (NM) from multiple sclerosis. Clone mCCD does not cause redistribution of Aquaporin-4 and also it does not affect coaggregation astrocytic CD32-B and Aquaporin-4. (Ref.: Hinson, SR et al. (2017). Proc. Natl. Acad. Sci. USA 114(21); 5491-5496).

Specificity

This rabbit polyclonal antiody detects Aquaporin-4. It targets an epitope with in 75 amino acids from the C-terminal.

Immunogen

Epitope: C-terminus
GST/HIs-taged recombinant fragment corresponding to 75 amino acids from the C-terminal region of rat Aquaporin-4.

Application

Anti-Aquaporin 4, Cat. No. ABN910, is a highly specific rabbit polyclonal antibody that targets Aquaporin 4 and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry (Paraffin) Analysis : A 1:1,000 dilution from a representative lot detected Aquaporin 4 in rat cerebral cortex tissue.

Western Blotting Analysis: A 1:500 dilution from a representative lot detected Aquaporin 4 in human brain tissue lysate.
Research Category
Neuroscience

Quality

Evaluated by Immunohistochemistry (Paraffin) in mouse cerebral cortex tissue.

Immunohistochemistry (Paraffin) Analysis : A 1:1,000 dilution of this antibody detected Aquaporin 4 in mouse cerebral cortex tissue.

Target description

~34.0 kDa observed; 34.48 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine, 150 mM NaCl (pH 7.4) with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nacoya Madden et al.
Frontiers in neuroscience, 17, 1198219-1198219 (2023-07-24)
The pathological involvement of the central nervous system in SARS-CoV2 (COVID-19) patients is established. The burden of pathology is most pronounced in the brain stem including the medulla oblongata. Hypoxic/ischemic damage is the most frequent neuropathologic abnormality. Other neuropathologic features
Rachel Britton et al.
Brain and behavior, 12(9), e2736-e2736 (2022-08-17)
Increasing age is the number one risk factor for developing cognitive decline and neurodegenerative disease. Aged humans and mice exhibit numerous molecular changes that contribute to a decline in cognitive function and increased risk of developing age-associated diseases. Here, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service