Skip to Content
Merck
All Photos(1)

Key Documents

ABD51

Sigma-Aldrich

Anti-BMPR-1A Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Bone morphogenetic protein receptor type-1A, BMP type-1A receptor, BMPR-1A, Activin receptor-like kinase 3, ALK-3, Serine/threonine-protein kinase receptor R5, SKR5, CD_antigen=CD292

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, human, mouse

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... BMPR1A(657)

General description

BMPR-1A (bone morphogenic protein receptor type 1A) signaling is known to be important for inner root sheath differentiation, hair follicle cycling and tooth development. Mutations in BMPR-1A are a common cause of juvenile polyposis syndrome (JPS). BMPR-1A is expressed in a high quantity during the formation of the primary and secondary palate and the loss of BMPR-1A could be a cause for the cleft palate birth defect.

Specificity

This antibody recognizes BMPR-1A at the extracellular domain.

Immunogen

Epitope: Extracellular domain
KLH-conjugated linear peptide corresponding to the extracellular domain of human BMPR-1A.

Application

Anti-BMPR-1A Antibody is an antibody against BMPR-1A for use in WB.
Research Category
Stem Cell Research
Research Sub Category
Kinases & Phosphatases

Quality

Evaluated by Western Blot in NIH/3T3 cell lysate.

Western Blot Analysis: 0.1 µg/mL of this antibody detected BMPR-1A on 10 µg of NIH/3T3 cell lysate.

Target description

~75 kDa observed. The calculated molecular weight is 67 kDa; however, BMPR-1A has been shown as a ~77 kDa band in western blots (Koenig, B., et al. (1994). Molecular and Cellular Biology. 14(9):5961-5974).

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
NIH/3T3 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joohwee Kim et al.
Biochimica et biophysica acta, 1853(2), 500-512 (2014-12-17)
Prostaglandins are a group of lipid signaling molecules involved in various physiological processes. In addition, prostaglandins have been implicated in the development and progression of diseases including cancer, cardiovascular disease, and arthritis. Prostaglandins exert their effects through the activation of
Guillermo Agustín Videla Richardson et al.
Brain pathology (Zurich, Switzerland), 26(1), 43-61 (2015-03-27)
Although BMP4-induced differentiation of glioma stem cells (GSCs) is well recognized, details of the cellular responses triggered by this morphogen are still poorly defined. In this study, we established several GSC-enriched cell lines (GSC-ECLs) from high-grade gliomas. The expansion of
Xing-Gang Wu et al.
Blood, 124(8), 1335-1343 (2014-06-07)
Mutations in HFE are the most common cause of hereditary hemochromatosis (HH). HFE mutations result in reduced expression of hepcidin, a hepatic hormone, which negatively regulates iron absorption from the duodenum and iron release from macrophages. However, the mechanism by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service