Skip to Content
Merck
All Photos(1)

Documents

AB5330

Sigma-Aldrich

Anti-Syntaxin 4 Antibody

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... STX4(6810)

Related Categories

Specificity

Recognizes Syntaxin 4.

Immunogen

Highly purified corresponding to residues 2-23 of rat or mouse Syntaxin 4 (Accession Q08850).

Application

Research Category
Neuroscience
Research Sub Category
Synapse & Synaptic Biology
This Anti-Syntaxin 4 Antibody is validated for use in WB for the detection of Syntaxin 4.
Western blotting: 0.5-1.0 μg/mL (1:200-1:500) Dilutions should be made using a carrier protein such as BSA (1-3%).


Optimal working dilutions must be determined by the end user.

Physical form

Affinity purified immunoglobulin. Lyophilized from PBS, pH 7.4, containing 1% BSA and 0.025% sodium azide. Reconstitute with 200 μL of sterile distilled water. Centrifuge antibody preparation before use (10,000 x g for 5 min).

Storage and Stability

Maintain lyophilized material at -20°C for up to 6 months. After reconstitution maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Analysis Note

Control
CONTROL ANTIGEN: Included free of charge with the antibody is 40 μg of control antigen (lyophilized powder). Reconstitute with 100 μL of sterile distilled water. For negative control, preincubate 1 μg of purified peptide with 1 μg of antibody for one hour at room temperature. Optimal concentrations must be determined by the end user.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arlene A Hirano et al.
Visual neuroscience, 24(4), 489-502 (2007-07-21)
Horizontal cells mediate inhibitory feed-forward and feedback communication in the outer retina; however, mechanisms that underlie transmitter release from mammalian horizontal cells are poorly understood. Toward determining whether the molecular machinery for exocytosis is present in horizontal cells, we investigated
Christian Puller et al.
Journal of the Optical Society of America. A, Optics, image science, and vision, 31(4), A189-A194 (2014-04-04)
The distribution of the soluble NSF-attachment protein receptor protein syntaxin-4 and the Na-K-Cl cotransporter (NKCC) were investigated in the outer plexiform layer of human retina using immunohistochemistry. Both proteins, which are proposed to be components of a gamma-aminobutyric acid mediated
Christian Puller et al.
PloS one, 9(2), e88963-e88963 (2014-03-04)
The functional roles and synaptic features of horizontal cells in the mammalian retina are still controversial. Evidence exists for feedback signaling from horizontal cells to cones and feed-forward signaling from horizontal cells to bipolar cells, but the details of the
Brendan J O'Brien et al.
Molecular vision, 16, 1854-1863 (2010-10-30)
The retina has the demanding task of encoding all aspects of the visual scene within the space of one fixation period lasting only a few hundred milliseconds. To accomplish this feat, information is encoded in specialized parallel channels and passed
Brian S Clark et al.
Development (Cambridge, England), 139(9), 1599-1610 (2012-04-12)
To gain insights into the cellular mechanisms of neurogenesis, we analyzed retinal neuroepithelia deficient for Llgl1, a protein implicated in apicobasal cell polarity, asymmetric cell division, cell shape and cell cycle exit. We found that vertebrate retinal neuroepithelia deficient for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service