Skip to Content
Merck
All Photos(2)

Documents

AB1607

Sigma-Aldrich

Anti-Phosphothreonine Antibody

Chemicon®, from rabbit

Synonym(s):

Phospho-Threonine Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity (predicted by homology)

all

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable
western blot: suitable

shipped in

wet ice

target post-translational modification

phosphorylation (pThr)

General description

Protein phosphorylation and dephosphorylation are basic mechanisms for the modification of protein function in eukaryotic cells. Phosphorylation is a rare post-translational event in normal tissue, however, the abundance of phosphorylated cellular proteins increases several fold following various activation processes which are mediated through phosphotyrosine, phosphoserine or phosphothreonine (p-tyr/p-ser/p-thr). Many signal transduction pathways, such as the EGF, PDGF and insulin receptor systems, contain tyr/ser/thr kinase which phosphorylate specific tyr/ser/thr residues upon binding of ligands to their receptors. T cell antigen receptor complex or the receptors for some hemopoietic growth factors may stimulate these phosphorylation associated kinases, and cells transformed by viral oncogenes contain elevated levels of phosphorylated tyr/ser/thr.

Specificity

Anti-phosphothreonine is species independent
By direct competitive ELISA methodology (antigen capture) the specificity of this antibody has been determined to be phosphothreonine. Binding of Ly-pT-Gly-HRP (0.2μg/100μL/well) to an anti-phosphothreonine coated microplate (0.5μg/100μL/well) is inhibited by 95% with the addition of 2-10 μg/mL anti-phosphothreonine. No inhibition is evident by inhibition of phosphoserine, phosphotyrosine, or threonine at concentrations of up to 1 mg/mL. Antibody AB1607 recognizes phosphothreonine, peptidyl-phosphothreonine, and threonine-phosphorylated proteins.

Immunogen

Keyhole Limpet Hemocyanin (KLH) – conjugated phosphothreonine conjugates and Phosvitin.

Application

Anti-Phosphothreonine Antibody is an antibody against Phosphothreonine for use in ELISA, IH(P), IP & WB.
ELISA (kinase assay):
A 0.5 μg/mL concentration of a previous lot was used in ELISA.

Immunoprecipitation:
A previous lot of this antibody was used in IP.

Immunohistochemistry (paraffin):
Representative image from a previous lot.

Optimal working dilutions must be determined by end user.
Research Category
Signaling
Research Sub Category
General Post-translation Modification

Quality

Routinely evaluated by Western Blot on Calyculin/Okadaic Acid treated HEK293 lysate.

Western Blot Analysis: 1:500 dilution of this lot detected PHOSPHOTHREONINE on 10 μg of Calyculin/Okadaic Acid treated HEK293 lysate.

Physical form

ImmunoAffinity Purified
Purified rabbit polyclonal in PBS buffer, containing 50% Glycerol, 0.01% NaN3.

Storage and Stability

Stable for 1 year at -20ºC in undiluted aliquots from date of receipt.

Handling Recommendations:
Upon receipt, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. Note: Variability in freezer temperatures below -20°C may cause glycerol-containing solutions to become frozen during storage.

Analysis Note

Control
Mouse brain lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yang Yang et al.
Frontiers in genetics, 13, 941171-941171 (2022-07-09)
BACKGROUND: Schizophrenia is a severe mental disorder with high heritability, and cognitive dysfunction is one of the core features. Growing evidence suggests the genetic risk of schizophrenia may contribute to cognitive impairments. The variant rs1635 (nucleotide sequence: c.455C>A; amino acid
Randa Abdel Malik et al.
Circulation research, 120(1), 99-109 (2016-10-26)
The AMP-activated protein kinase (AMPK) is stimulated by hypoxia, and although the AMPKα1 catalytic subunit has been implicated in angiogenesis, little is known about the role played by the AMPKα2 subunit in vascular repair. To determine the role of the
Brian D Carey et al.
PLoS pathogens, 16(3), e1008282-e1008282 (2020-03-10)
Protein phosphorylation plays an important role during the life cycle of many viruses. Venezuelan equine encephalitis virus (VEEV) capsid protein has recently been shown to be phosphorylated at four residues. Here those studies are extended to determine the kinase responsible
Brian D Carey et al.
Journal of virology, 92(15) (2018-05-18)
Protein phosphatase 1 (PP1) is a serine/threonine phosphatase which has been implicated in the regulation of a number of viruses, including HIV-1, Ebolavirus, and Rift Valley fever virus. Catalytic subunits of PP1 (PP1α, PP1β, and PP1γ) interact with a host
Mercy Pawar et al.
Cell death and differentiation, 24(10), 1799-1810 (2017-07-15)
We report the neuroprotective role of FAS apoptotic inhibitory molecule 2 (FAIM2), an inhibitor of the FAS signaling pathway, during stress-induced photoreceptor apoptosis. Retinal detachment resulted in increased FAIM2 levels in photoreceptors with higher amounts detected at the tips of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service