No, 70% alcohol (product 65350-85) should not be used as a disinfectant. If the product were pure 70% ethanol, then perhaps it could be used as a disinfectant. However, this particular 70% alcohol is actually a reagent grade alcohol, referred to as Specially Denatured 3A or SD3A. The composition is approximately 63-64% ethanol, and it also contains about 4% each of isopropyl alcohol and methanol. It's important to note that methanol is a poison and is readily absorbed through the skin. The body converts the methanol to formaldehyde, and each year numerous people worldwide die from methanol poisoning. Instead of using Reagent Alcohol, it would be best to use either 70% pure ethanol or 70% pure isopropyl alcohol.
65350-M
Alcohol 70%
HARLECO® Used to hydrate or dehydrate tissue samples in tissue processing and staining protocols
Synonym(s):
Ethanol solution, Dehydration, Ethyl alcohol
About This Item
hematology
histology
Recommended Products
Quality Level
form
liquid
transition temp
flash point 12 °C (Information refers to the main component.)
application(s)
diagnostic assay manufacturing
hematology
histology
storage temp.
10-30°C
SMILES string
CCO
InChI
1S/C2H6O/c1-2-3/h3H,2H2,1H3
InChI key
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 2
Target Organs
Eyes,Central nervous system
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
53.6 °F - Information refers to the main ingredient.
Flash Point(C)
12 °C - Information refers to the main ingredient.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
Can 70% alcohol (65350-85) be used as a disinfectant?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service