Skip to Content
Merck
All Photos(1)

Key Documents

5.33978

Sigma-Aldrich

ACC Inhibitor IV, CP-640186

Synonym(s):

ACC Inhibitor IV, CP-640186, 9-Anthryl((3 R)-3-(4-morpholinylcarbonyl)-1,4ʹ-bipiperidin-1ʹ-yl)methanone, HCl, Acetyl-CoA Carboxylase Inhibitor IV, CP-640186, HCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H35N3O3
Molecular Weight:
485.62
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

light orange

solubility

DMSO: 50 mg/mL
H2O: 50 mg/mL

storage temp.

2-8°C

General description

A cell-permeable, orally active, non-toxic bipiperidylcarboxamide derived compound that binds to the active site of acetyl-CoA carboxylase (ACC) at the interface between the two monomers of the CT dimer and acts as a potent, allosteric, and reversible, inhibitor of ACC (IC50 = 53 nM and 61 nM for rat liver ACC1 and ACC2). The inhibition is uncompetitive with respect to ATP and non-competitive with respect to acetyl-CoA, citrate, and bicarbonate. Shown to inhibit fatty acid synthesis in Sprague-Dawley rats, and in CD1 and ob/ob mice (ED50 = 13, 11, and 4 mg/kg). Stimulates fatty acid oxidation in CC12 cells in rat epitrochelearis muscle (ED50 = 57 nM) and reduces muscle malonyl-CoA levels. Improves insulin sensitivity in diet-induced obese animal models. Displays good metabolic stability and moderate pharmacokinetic properties (plasma half-life =1.5 h; Cmax = 345 ng/ml; and AUC = 960 ng/h/ml at 5 mg/kg i.v. or 10 mg/kg p.o.).
ACC Inhibitor IV, CP-640186, is a cell-permeable, potent, allosteric, eversible inhibitor of ACC (IC₅₀ = 53 and 61 nM for rat liver ACC1 & ACC2). The inhibition is uncompetitive with respect to ATP.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Acetyl-CoA carboxylase (ACC)
Target IC50: 53 nM and 61 nM for rat liver ACC1 and ACC2

Warning

Toxicity: Standard Handling (A)

Physical form

Supplied as a hydrochloride salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Tong, L., and Harwood, H.J., 2006. J. Cell. Biol.99, 1476.
Gu, Y.G., et al. 2006. J. Med. Chem.49, 3770.

Zhang, H., et al. 2004. Structure12, 1683.

Harwood, H.J., et al. 2003. J. Biol. Chem.278, 37099.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service