Skip to Content
Merck
All Photos(1)

Key Documents

528101

Sigma-Aldrich

PI-103

InSolution, ≥97%, potent ATP-competitive inhibitor of DNA-PK, PI3-K, and mTOR

Synonym(s):

InSolution PI-103, mTOR Inhibitor V, PI 3-K Inhibitor V

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H16N4O3
Molecular Weight:
348.36
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

shipped in

wet ice

storage temp.

−20°C

General description

A cell-permeable pyridinylfuranopyrimidine compound that acts as a potent and ATP-competitive inhibitor of DNA-PK, PI3-K, and mTOR (IC50 = 2, 8, 88, 48, 150, 26, 20, and 83 nM for DNA-PK, p110α, p110β, p110δ, p110γ, PI3-KC2β, mTORC1, and mTORC2, respectively). It inhibits ATR and ATM only at much higher concentrations (IC50 = 850 and 920 nM, respectively) and exhibits little activity towards a panel of more than 40 other kinases even at concentrations as high as 10 µM. Shown to effectively block PI3-K/Akt signaling and cell proliferation in glioma cell lines both in vitro and in vivo.

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Physical form

A 10 mM (2 mg/574 µl) solution of PI-103 (Cat. No. 528100) in DMSO.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Raynaud, F.I., et al. 2007. Cancer Res.67, 5840.
Fan, Q.W., et al. 2006. Cancer Cell9, 341.
Knight, Z.A., et al. 2006. Cell125, 733.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F - closed cup - (Dimethylsulfoxide)

Flash Point(C)

87 °C - closed cup - (Dimethylsulfoxide)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service