Skip to Content
Merck
All Photos(1)

Key Documents

5.09267

Sigma-Aldrich

Opn4 Antagonist

Synonym(s):

Opn4 Antagonist, Opn4 Antagonist, Opsinamide, Melanopsin-inhibiting Sulfonamide AA92593, 1-(4-Methoxy-3-methyl-benzenesulfonyl)piperidine, 1-(4-Methoxy-3-methylphenylsulfonyl)piperidine, Melanopsin Antagonist, Opsinamide AA92593

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H19NO3S
CAS Number:
Molecular Weight:
269.36
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(N2CCCCC2)c1cc(c(cc1)OC)C

InChI

1S/C13H19NO3S/c1-11-10-12(6-7-13(11)17-2)18(15,16)14-8-4-3-5-9-14/h6-7,10H,3-5,8-9H2,1-2H3

InChI key

HDTKLZINZGEPFG-UHFFFAOYSA-N

General description

A sulfonamide compound that prevents melanopsin/Opn4 photoactivation in a reversible and cis-Retinal-competitive manner (Kb = 6 and 0.16 µM in competitive binding studies using Opn4 pre-bound with cis-Retinal or not) without apparent affinity toward bovine retina rhodopsin or a panel of 74 other receptors, ion channels, and enzymes. Effectively inhibits cellular phototransduction mediated by human Opn4 in CHO transfectants (IC50 = 665 nM; drug added 30 min before 10 µM 9-cis-Retinal & photoactivation), by murine Opn4 in transfected Xenopus oocytes (IC50 = 190 nM; drug added 3 min before 50 µM 11-cis-Retinal and photoactivation), as well as by endogenous Opn4 in primary rat ipRGCs (intrinsically photosensitive retinal ganglion cells; 10 µM). Shown to be bioavailable in mice via i.p. injection (retina [AA92593] ~7.5 µM 30 min post 30 mg/kg i.p. dosing; >95% clearance within 2 h) and effectively suppress PLR (pupillary light reflex; pupil constriction) in a time- and Opn4-dependent manner in mice in vivo. Likewise, blue light insensitivity is observed in Opn4-/- and AA92593-treated wild-type, but not vehicle-treated wild-type, P14 neonatal mice.
A sulfonamide compound that prevents melanopsin/Opn4 photoactivation in a reversible and cis-Retinal-competitive manner without apparent affinity toward bovine retina rhodopsin or a panel of 74 other receptors, ion channels, and enzymes. Effectively inhibits cellular phototransduction mediated by human Opn4 (IC50 = 665 nM; using CHO transfectants), murine Opn4 (IC50 = 190 nM; using transfected Xenopus oocytes), and rat Opn4 (10 µM; using primary rat ipRGCs). Shown to effectively suppress PLR (pupillary light reflex; pupil constriction) in a time- and Opn4-dependent manner in mice (30 mg/kg i.p.) in vivo. Likewise, blue light insensitivity is observed in Opn4-/- and AA92593-treated wild-type, but not vehicle-treated wild-type, P14 neonatal mice.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
melanopsin (Opn4)
Reversible: yes
Target Ki: 110 and 160 nM for human and mouse melanopsin, respectively)

Packaging

Packaged under inert gas

Warning

Toxicity: Regulatory Review (Z)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 monthst at -20°C.

Other Notes

Jones, K.A., et al. 2013. Nat. Chem. Biol.9, 630.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service