Skip to Content
Merck
All Photos(1)

Key Documents

474422

Sigma-Aldrich

4-Methylumbelliferyl-α-D-galactopyranoside

Substrate for the fluorogenic assay of α-D-galactosidase.

Synonym(s):

4-Methylumbelliferyl-α-D-galactopyranoside, 4-MU-α-D-Gal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H18O8
CAS Number:
Molecular Weight:
338.31
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated
protect from light

impurities

≤100 ppm 4-methylumbelliferone

color

white to off-white

solubility

DMF: 50 mg/mL

fluorescence

λex 360 nm
λem 440 nm

shipped in

ambient

storage temp.

−20°C

SMILES string

O1C(C(C(C(C1CO)O)O)O)Oc2cc3[o][c](cc(c3cc2)C)=O

InChI

1S/C16H18O8/c1-7-4-12(18)23-10-5-8(2-3-9(7)10)22-16-15(21)14(20)13(19)11(6-17)24-16/h2-5,11,13-17,19-21H,6H2,1H3

InChI key

YUDPTGPSBJVHCN-UHFFFAOYSA-N

General description

Substrate for the fluorogenic assay of α-D-galactosidase. Excitation maximum: ~360 nm; Emission maximum: ~440 nm.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Unstable in solution; reconstitute just prior to use. Solubilization in DMF is preferred, as background fluorescence will increase if solubilized in H₂O.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service