Skip to Content
Merck
All Photos(1)

Key Documents

459618

Sigma-Aldrich

Okadaic Acid potassium salt

≥95% (HPLC), solid, protein phosphatase 2A and protein phosphatase 1 inhibitor, Calbiochem®

Synonym(s):

Okadaic Acid, Potassium Salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H67O13 · K
CAS Number:
Molecular Weight:
843.09
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.77

Product Name

Okadaic Acid, Potassium Salt, Water-soluble analog of Okadaic Acid. Inhibits protein phosphatases 1 and 2A.

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

water: 1 mg/mL
DMSO: 20 mg/mL
ethanol: 20 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

[K+].[O-]C(=O)C(O)(CC1OC2(OC(CC(=C2)C)C(C)C=CC3OC4(OC5C(OC(C(=C)C5O)C(O)CC(C6OC7(OCCCC7)CCC6C)C)CC4)CC3)C(CC1)O)C

InChI

1S/C44H68O13.K/c1-25-21-34(55-44(23-25)35(46)12-11-31(54-44)24-41(6,50)40(48)49)26(2)9-10-30-14-18-43(53-30)19-15-33-39(57-43)36(47)29(5)38(52-33)32(45)22-28(4)37-27(3)13-17-42(56-37)16-7-8-20-51-42;/h9-10,23,26-28,30-39,45-47,50H,5,7-8,11-22,24H2,1-4,6H3,(H,48,49);/q;+1/p-1

InChI key

UXRQUXBFVICHQJ-UHFFFAOYSA-M

General description

Water-soluble analog of Okadaic Acid (Cat. No. 495604). Inhibits protein phosphatases 1 and 2A.

Biochem/physiol Actions

Cell permeable: no
Primary Target
PP1, PP2
Product does not compete with ATP.
Reversible: no
Target IC50: 10-15 nM, 0.1 nM, against protein phosphatase 1 and protein phosphatase 2A, respectively

Warning

Toxicity: Toxic (F)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Gjertsen, B.T., et al. 1994. J. Cell Sci.107, 3363.
Kiguchi, K., et al. 1994. Cell Growth Differ.5, 995.
Ohaka, Y., et al. 1993. Biochem. Biophys. Res. Commun.197, 916.
Gopalakrishna, R., et al. 1992. Biochem. Biophys. Res. Commun.189, 950.
Kreienbuhl, P., et al. 1992. Blood80, 2911.
Nomura, M., et al. 1992. Biochemistry31, 11915.
Song, Q., et al. 1992. J. Cell Physiol.153, 550.
Tada, Y., et al. 1992. Immunopharmacol.24, 17.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Irene Benito-Cuesta et al.
Autophagy, 17(3), 656-671 (2020-02-23)
The physiological AKT-MTORC1 and AMPK signaling pathways are considered key nodes in the regulation of anabolism-catabolism, and particularly of macroautophagy/autophagy. Indeed, it is reported that these are altered processes in neurodegenerative proteinopathies such as Alzheimer disease (AD), mainly characterized by
Andrés Vicente-Acosta et al.
Journal of neuroinflammation, 19(1), 93-93 (2022-04-14)
Friedreich's ataxia is a rare hereditary neurodegenerative disease caused by decreased levels of the mitochondrial protein frataxin. Similar to other neurodegenerative pathologies, previous studies suggested that astrocytes might contribute to the progression of the disease. To fully understand the mechanisms

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service