Skip to Content
Merck
All Photos(1)

Key Documents

375054

Millipore

Heparin, Porcine Intestinal Mucosa, Biotin Conjugate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352201
NACRES:
NA.79

biological source

Porcine intestinal mucosa

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

solubility

water: soluble

shipped in

ambient

storage temp.

2-8°C

General description

Biotinylated heparin containing 5-8 mole-percent biotin. Biotin attachment is through a 12-atom spacer arm containing a cleavable disulfide group.
Biotinylated heparin containing 5-8 mole-percent biotin. Biotin attachment is through a 12-atom spacer arm containing a cleavable disulfide group. M.W. 15000 (avg.).

Warning

Toxicity: Harmful (C)

Other Notes

Pankhurst, G.J., et al. 1998. Biochemistry 37, 4823.
Stearns, N.A., et al. 1997. Anal. Biochem. 247, 348.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service