Skip to Content
Merck

20-145

Assay Dilution Buffer, 5X

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41116012
NACRES:
NA.41
eCl@ss:
32160405
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

liquid

Quality Segment

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

shipped in

wet ice

Application

For use in kinase assays.
Research Category
All

Physical form

100mM MOPS, pH 7.2, 125mM β-glycerophosphate, 25mM EGTA, 5mM sodium orthovanadate, 5mM dithiothreitol

Preparation Note

6 months at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library