Skip to Content
Merck
All Photos(1)

Key Documents

1.93415

Supelco

Formic acid

98-100%, EMPLURA®

Synonym(s):

Formic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCOOH
CAS Number:
Molecular Weight:
46.03
Beilstein:
1209246
MDL number:
UNSPSC Code:
12352106

vapor density

1.6 (vs air)

vapor pressure

44.8 mmHg ( 20 °C)

product line

EMPLURA®

Assay

98-100%

autoignition temp.

1004 °F

expl. lim.

57 %

refractive index

n20/D 1.370 (lit.)

bp

100-101 °C (lit.)

mp

8.2-8.4 °C (lit.)

density

1.22 g/mL at 25 °C (lit.)

storage temp.

room temp

SMILES string

OC=O

InChI

1S/CH2O2/c2-1-3/h1H,(H,2,3)

InChI key

BDAGIHXWWSANSR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

1.93415.0521: 500 mL, Minimum order quantity: 20
1.93415.9026: 25 L, Minimum order quantity: 1

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

121.1 °F - closed cup

Flash Point(C)

49.5 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Inorganics and solvents for basic and standard applications manufactured locally.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service