Skip to Content
Merck
All Photos(1)

Key Documents

1191

Millipore

Adenosine 5′-Triphosphate, Disodium Salt

Synonym(s):

Adenosine 5′-Triphosphate, Disodium Salt, ATP, 2Na

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14N5O13P3 · 2Na
CAS Number:
Molecular Weight:
551.14
MDL number:
UNSPSC Code:
12352208
NACRES:
NA.51

description

RTECS - AU7417000
Merck USA index - 14, 155

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

water: 20 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

[Na+].[Na+].[P](=O)([O-])(O[P](=O)(O[P](=O)([O-])O)O)OC[C@H]1O[C@H]([C@@H]([C@@H]1O)O)[n]2c3ncnc(c3nc2)N

InChI

1S/C10H16N5O13P3.2Na/c11-8-5-9(13-2-12-8)15(3-14-5)10-7(17)6(16)4(26-10)1-25-30(21,22)28-31(23,24)27-29(18,19)20;;/h2-4,6-7,10,16-17H,1H2,(H,21,22)(H,23,24)(H2,11,12,13)(H2,18,19,20);;/q;2*+1/p-2/t4-,6-,7-,10-;;/m1../s1

InChI key

TTWYZDPBDWHJOR-IDIVVRGQSA-L

Related Categories

General description

Coenzyme involved in the transfer of phosphate bond energy.

Application


  • Asynchronising five-fold symmetry sequence for better homonuclear polarisation transfer in magic-angle-spinning solid-state NMR.: This study highlights the use of Adenosine 5′-Triphosphate, Disodium Salt (ATP) in enhancing nuclear magnetic resonance (NMR) spectroscopy techniques, which is pivotal in structural biochemistry and the study of molecular dynamics (Arunachalam et al., 2023).

  • Insights into the conversion of dissolved organic phosphorus favors algal bloom, arsenate biotransformation and microcystins release of Microcystis aeruginosa.: This research utilizes ATP as a phosphorus source in studies of algal blooms, offering significant insights into environmental biochemistry and the biotransformation processes that impact water quality and ecosystem health (Zhang et al., 2023).

  • Overview of Portable Assays for the Detection of Mycotoxins, Allergens, and Sanitation Monitoring.: ATP is utilized in the development of portable assays for rapid on-site testing in food safety and public health, demonstrating its versatility and essential role in biochemical assays aimed at improving global health outcomes (Sarver et al., 2021).

  • Organic Phosphorous and Calcium Source Induce the Synthesis of Yolk-Shell Structured Microspheres of Calcium Phosphate with High-Specific Surface Area: Application in HEL Adsorption.: This study illustrates the application of ATP in synthesizing advanced materials for biomedical applications, specifically in the development of high-surface-area microspheres for therapeutic protein adsorption (Cao et al., 2020).

  • Inflammasome-Driven Interleukin-1α and Interleukin-1β Production in Atherosclerotic Plaques Relates to Hyperlipidemia and Plaque Complexity.: ATP′s role is investigated in cellular signaling pathways involved in inflammation and cardiovascular disease, providing crucial insights into the pathophysiology of atherosclerosis and potential therapeutic targets (Jiang et al., 2019).


Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-70°C). Stock solutions are stable for up to 2-3 h at 4°C or for up to 3 months at -70°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service