Skip to Content
Merck
All Photos(1)

Key Documents

1.09495

Supelco

ICP multi-element standard solution XVII

(7 elements in hydrochloric acid 15%) 100 mg/l: Hf, Ir, Sb, Sn, Ta, Ti, Zr Certipur®

Synonym(s):

Multi-Element ICP Standard Solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

reference material

Quality Level

product line

Certipur®

technique(s)

ICP: suitable

pH

1 (20 °C in H2O)

density

1.076 g/cm3 at 20 °C

application(s)

industrial qc
pharmaceutical

format

multi-component solution

storage temp.

15-25°C

Analysis Note

Hf (Hafnium): 100 mg/l
Ir (Iridium): 100 mg/l
Sb (Antimony): 100 mg/l
Sn (Tin): 100 mg/l
Ta (Tantalum): 100 mg/l
Ti (Titanium): 100 mg/l
Zr (Zirconium): 100 mg/l

Other Notes

For a complete product listing of our ICP and AAS range, including technical information and example certificates, please visit ICP & AAS standards website.

Detailed information on our complete workflow solutions for ICP-AAS can be found in our Brochure: High-Purity Chemicals for Inorganic Trace Analysis.

In our effort to be more sustainable, we′re going digital with COAs-download them directly from our website.

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service