Skip to Content
Merck
All Photos(1)

Key Documents

1.08165

Supelco

UV-VIS Standard 5: Toluene solution in n-hexane

according to Ph. Eur. resolution power testing, Certipur®

Synonym(s):

UV-VIS Standard 5 - Toluene standard solution (0.02% in n-hexane)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Agency

according to Ph. Eur. resolution power testing

product line

Certipur®

technique(s)

UV/Vis spectroscopy: suitable

application(s)

environmental
industrial qc

format

mixture

storage temp.

15-25°C

Application

UV-VIS Standard 5: Toluene solution in n-hexane as a reference material for checking the spectral resolution of spectrophotometers in accordance with European Pharmacopoeia (Ph. Eur.) chapter 2.2.25.

Features and Benefits

  • It is a solution of 0.02 % toluene in n-hexane
  • Conveniently packed in 10 mL glass ampules. Each box includes 3 single-use ampules
  • Stable solutions in glass ampoules
  • Accreditation according to ISO 17025
  • Traceability to standard reference material from the National Institute of Standards and Technology (NIST) and Physical-Technical Institute (PTB)
  • Includes a certificate of analysis (CoA)
  • Reliable and reproducible results

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

3 - Flammable liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service