Skip to Content
Merck
All Photos(1)

Key Documents

1.06352

Supelco

Sodium hydrogen sulfate monohydrate

for analysis EMSURE®

Synonym(s):

Sodium hydrogen sulfate monohydrate, sodium bisulfate monohydrate, sodium bisulphate monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HNaO4S · xH2O
CAS Number:
Molecular Weight:
120.06 (anhydrous basis)
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-665-7
NACRES:
NA.21
Assay:
≥99.0% (alkalimetric)
form:
solid
solubility:
1080 g/L

Quality Level

product line

EMSURE®

Assay

≥99.0% (alkalimetric)

form

solid

potency

2490 mg/kg LD50, oral (Rat)

pH

1 (20 °C, 50 g/L in H2O)

mp

183 °C (decomposition)

solubility

1080 g/L

bulk density

900‑970 kg/m3

anion traces

chloride (Cl-): ≤0.001%
phosphate (PO43-): ≤0.0005%

cation traces

Al: ≤0.001%
Ca: ≤0.001%
Fe: ≤0.0005%
K: ≤0.002%
Mg: ≤0.0005%
N: ≤0.001%
heavy metals (as Pb): ≤0.0005%

storage temp.

2-30°C

SMILES string

[Na+].[S](=O)(=O)(O)O.[O-H]

InChI

1S/Na.H2O4S.H2O/c;1-5(2,3)4;/h;(H2,1,2,3,4);1H2/q+1;;/p-1

InChI key

JXHZRQHZVYDRGX-UHFFFAOYSA-M

Analysis Note

Assay (alkalimetric): ≥ 99.0 %
Chloride (Cl): ≤ 0.001 %
Phosphate (PO₄): ≤ 0.0005 %
Total nitrogen (N): ≤ 0.001 %
Heavy metals (as Pb): ≤ 0.0005 %
Al (Aluminium): ≤ 0.001 %
Ca (Calcium): ≤ 0.001 %
Fe (Iron): ≤ 0.0005 %
K (Potassium): ≤ 0.002 %
Mg (Magnesium): ≤ 0.0005 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service