1.03795
Cellvento® 4CHO COMP
chemically defined cell culture medium, ≤1.0 IU/mL endotoxin, pH 5.1-5.6(without supplement), pH 6.7-7.3(with supplement)
Pharma Manufacturing
About This Item
Quality Level
description
shipped with cooling packs
product line
Cellvento®
form
dry powder
technique(s)
cell culture | mammalian: suitable (suspension)
storage temp.
2-8°C
General description
Application
Quantity
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Sens. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
This case study walks through optimizing feed and media for upstream bioprocessing and how it translates to large-scale bioreactors.
Explore how low-impurity iron sources enhance consistency and control in recombinant protein production using Cellvento® 4CHO medium.
Explore how low-impurity iron sources enhance consistency and control in recombinant protein production using Cellvento® 4CHO medium.
Explore how low-impurity iron sources enhance consistency and control in recombinant protein production using Cellvento® 4CHO medium.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service