Skip to Content
Merck
All Photos(5)

Key Documents

07-145

Sigma-Aldrich

Anti-phospho-Histone H3 (Ser28) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Histone H3.1t, H3/t, H3t, H3/g

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse

technique(s)

ChIP: suitable
dot blot: suitable
immunocytochemistry: suitable
inhibition assay: suitable (peptide)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pSer28)

Gene Information

human ... H3C1(8350)
mouse ... H3C1(360198)

General description

Histone H3 is one of the five main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N-terminal tail, H3 is involved with the structure of the nucleosomes of the ′beads on a string′ structure. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of epigenetic modifications that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine.

Specificity

This antibody is expected to demonstrate broad species reactivity due to sequence conservation.
This antibody recognizes Histone H3 phosphorylated at Ser28.

Immunogen

Epitope: Histone H3 phosphorylated at Ser28
KLH-conjugated linear peptide corresponding to Histone H3 phosphorylated at Ser28.

Application

Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones
Use Anti-phospho-Histone H3 (Ser28) Antibody (Rabbit Polyclonal Antibody) validated in WB, ICC, DB, PIA, ChIP to detect phospho-Histone H3 (Ser28) also known as H3F3, H3F3B, H3S28P.
Western Blot Analysis: A 1:2,000 dilution from a representative lot detected phospho-Histone H3 (Ser28) in 10 µg of untreated and colcemid treated HeLa acid extract.

Immunocytochemistry Analysis: A 1:500 dilution from a representative lot detected phospho-Histone H3 (Ser28) in HeLa cells.

Chromatin Immunoprecipitation Analysis: A representative lot immunoprecipitated phospho-Histone H3 (Ser28) in colcecmid treated HeLa cell lysate.

Dot Blot Specificity Analysis: A representative lot of this antibody detected peptides corresponding to phospho-Histone H3 (Ser28), but not peptides corresponding to other unmodified and modifed Histones.

Peptide Inhibition Analysis: A 1:500 dilution form a representative was blocked by phospho-Histone H3 (Ser28) peptides, but not non-specific peptides.

Quality

Evaluated by Western Blot in HeLa acid extract.

Western Blot Analysis: A 1:2,000 dilution of this antibody detected phospho-Histone H3 (Ser28) in 10 µg of HeLa acid extract.

Target description

~17 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa acid extract

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dazl deficiency leads to embryonic arrest of germ cell development in XY C57BL/6 mice.
Lin, Y; Page, DC
Developmental Biology null
G1 phase regulation, area-specific cell cycle control, and cytoarchitectonics in the primate cortex.
Lukaszewicz, A; Savatier, P; Cortay, V; Giroud, P; Huissoud, C; Berland, M; Kennedy, H; Dehay, C
Neuron null
Topoisomerase inhibitor induced dephosphorylation of H1 and H3 histones as a consequence of cell cycle arrest.
Nicole Happel, Arne Sommer, Kristina Hanecke, Werner Albig, Detlef Doenecke
Journal of Cellular Biochemistry null
Role of apoptosis in low-dose hyper-radiosensitivity.
S A Krueger,M C Joiner,M Weinfeld,E Piasentin,B Marples
Radiation Research null
Adrien Demilly et al.
PloS one, 6(6), e21213-e21213 (2011-06-23)
Genes of the coe (collier/olfactory/early B-cell factor) family encode Helix-Loop-Helix transcription factors that are widely conserved in metazoans and involved in many developmental processes, neurogenesis in particular. Whereas their functions during vertebrate neural tube formation have been well documented, very

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service