Key Documents
Z687995
Plas-Labs temperature and humidity control box
AC/DC input 240 V AC, UK plug, Plas-Labs 890-THC/EXP/UK
About This Item
UNSPSC Code:
41123304
NACRES:
NB.35
feature
CE compliant
manufacturer/tradename
Plas-Labs 890-THC/EXP/UK
parameter
16-44 °C temp. range
inside H × W × D
26 in. × 41 in. × 28 in.
outside H × W × D
38 in. × 55 in. × 35 in.
General description
An inert gas source is required for humidity control. One piece clear acrylic formed top, white thermoset bottom with easy to clean corners, and one vacuum capable transfer chamber (12 in. L x 11 in. I.D) with two (2) gas purging valves and one vacuum gauge.
The 8 inch color touchscreen features: one-touch on-screen 24 h datalogging, real time clock, audible alarms, four preset conditions, separate on/off control for temperature
and humidity, and password protected administration window for PID adjustment, test I/O, real time clock adjustment, and alarm conditions.
  • 8 inch color touch screen display
  • Thermoelectric(Peltier) assembly
  • Nebulizer
  • Inert gas hook-up
  • One pair of white Hypalon ambidextrous gloves
  • Two (2) ground key cock valves for purging
  • One hospital grade multiple electric outlet strip
  • Fluorescent light and serial port connection fordatalogging.
  • Humidity range: 5-95% Rh
Legal Information
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)