Skip to Content
Merck
All Photos(1)

Key Documents

Z562963

Plas-Labs compact glove box

AC/DC input 120 V AC, Plas-Labs 830-ABC

Synonym(s):

humidity chambers, humidity control chambers, isolation chambers, nitrogen dry box

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103406

manufacturer/tradename

Plas-Labs 830-ABC

inside W × D × H

28 in. × 23 in. × 29 in.

outside W × D × H

43 in. × 24 in. × 31 in.

Looking for similar products? Visit Product Comparison Guide

General description

The Plas-Labs compact glove box features cleanliness, safety, complete containment, porability, and a draft-free atmosphere. The tall profile has a small footprint and is especially suited for hazardous materials to be weighed.

The compact glove box chamber is ideal for determining the dry weight or moisture content of aqueous solutions, adhesives, cereals, toxic chemicals, pharmaceuticals, paper, plastics, and light radioactive materials. It minimizes product degradation caused by moisture absorption.

The compact glove box may also be used with analytical balances, top loaders, bench scales and moisture analyzers, and is suitable for handling toxic materials and pharmaceuticals.

  • Removable top section for easy installation of equipment
  • Adjustable SS clamp
  • Ambidextrous Hypalon® gloves with SS O-rings
  • Transparent transfer chamber (12 L × 11 in. I.D.) with adjustable vacuum gauge
  • White levelling tray for transferring liquids through the transfer chamber
  • Four purge valves, two on transfer chamber, two on main chamber
  • HEPA and organic vapour canister filters (replaceable)
  • Multiple electrical outlet strip, CE UL and CSA approved

Legal Information

Hypalon is a registered trademark of E. I. du Pont de Nemours and Company

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service