Key Documents
Z549703
Sigma-Aldrich
High-vacuum valve adapter
male joint: ST/NS 24/40
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
Joint
male joint: ST/NS24/40
Related Categories
General description
For high-vacuum applications. Features a high-vacuum PTFE valve. For use with 1/4in. I.D. tubing.
Choose from one of the most recent versions:
Certificates of Analysis (COA)