Z508470
Ace flat-bottom reaction flasks
capacity 4,000 mL, w/o O-ring groove
Synonym(s):
bottom flask, flat flask, reaction flasks
All Photos(3)
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
Recommended Products
material
flat bottom
glass
single-neck
sterility
non-sterile
feature
closure type Lid
packaging
pack of 1
manufacturer/tradename
Ace Glass 6511-31
capacity
4,000 mL
Related Categories
General description
Cylindrical heavy-wall reaction flask with a flat bottom rounded into side of wall, and extra-thick flat flange. No constriction at top facilitates introduction or removal of material and allows easy cleaning. Optional O-ring groove allows use of CAPFE O-rings rather than gaskets. Flask is supplied with one CAPFE O-ring. Order reaction flask heads and clamps separately.
Choose from one of the most recent versions:
Certificates of Analysis (COA)