WHA10372803
Whatman® glass microfiber grade GF 31 1.4 µm pore size
white, binder free, 100 ea, 55 mm diam
Synonym(s):
Z750131
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
40161507
NACRES:
NB.24
Recommended Products
product name
Whatman® glass microfiber filters, Grade 31, diam. 55 mm
material
borosilicate glass membrane
circles
packaging
pk of 100
manufacturer/tradename
Whatman 10372803
Whatman Article No. 28414828 (US reference)
diam.
55 mm
Other Notes
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Legal Information
Whatman is a registered trademark of Cytiva
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service