Skip to Content
Merck
All Photos(1)

Key Documents

WAMUPG2MCPWC8

Whatman® Mini-UniPrep® G2 hand compressor

8 position, pack of 1

Synonym(s):

hand compressor, mini-uniprep G2, syringeless filter, whatman filter

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40151600
NACRES:
NB.24

packaging

pack of 1

manufacturer/tradename

Whatman Article No. 29026196 (US reference)
Whatman MUPG2MCPWC8

Looking for similar products? Visit Product Comparison Guide

General description

Mini-UniPrep G2 Hand Compressor is for use with Mini-UniPrep G2 Syringeless filters.

Features and Benefits

  • Streamlines syringeless filtration by filtering up to eight filter vials simultaneously
  • Designed for Cytiva′s Mini-UniPrepG2 - with integral borosilicate glass autosampler vial, plunger and attached filter membrane, and septum with cap
  • Glass construction prevents interference of leachables from plastic housing with HPLC/UHPLC pharmaceutical analysis
  • Mini-UniPrepG2 vials can be loaded directly into the autosampler

Other Notes

Contains a Multicompressor and a single 8 position white tray to be used for glass MUP G2

Legal Information

Mini-UniPrep is a registered trademark of Cytiva
Whatman is a registered trademark of Cytiva

recommended


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service