TEM-LC300CU100
Lacey Carbon Supported Copper Grids
size 300 mesh, size 100 nm , hole size, box of 25 ×
Synonym(s):
Lacey/ Carbon Film - Cu, 300 mesh 100 micron, 25pk
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
Recommended Products
packaging
box of 25 ×
size
100 nm , hole size
300 mesh
Looking for similar products? Visit Product Comparison Guide
General description
Lacey carbon film supported TEM grids. This carbon coating consists of woven-mesh-like openings of different sizes and shapes. This type of pattern provides support but does not interfere when observing specimen sections.
Application
- Investigation of Carbon Support on Ruthenium/Catalysts: This study explores the impact of carbon support properties on the efficiency of ruthenium/carbon catalysts for hydrogenation processes, using lacey carbon films on copper grids for catalyst characterization (Jones et al., 2016).
- Nitrogen Doping of Carbon Supports for Hydrogen Production: Analyzes the effects of nitrogen-doped carbon supports for nickel catalysts in hydrogen production from formic acid, with characterizations performed on lacey carbon supported copper grids (Nishchakova et al., 2019).
- Removal of Uranium on Carbon-Supported Iron Particles: Discusses the use of carbon-supported nanoscale zero-valent iron particles for uranium removal, where samples were mounted on lacey carbon supported copper grids for analysis (Crane & Scott, 2014).
- Redispersion of Carbon-Supported Metal Catalysts: Focuses on the redispersion strategies for high-loading metal catalysts on carbon supports, tested using lacey carbon supported copper grids for catalyst characterization (Giulimondi et al., 2022).
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service