Key Documents
HAM201100
Hamilton® Gastight® syringe, general use, 1700 series
model, 1725, AccuDil (AD) end, volume 250 μL
About This Item
UNSPSC Code:
41122001
NACRES:
NB.03
material
Borosilicate glass/PTFE
Gastight syringe
feature
autoclavable: no
model 1725, AccuDil (AD) end
manufacturer/tradename
Hamilton
parameter
20-50 °C temp. range (68-122 °F)
500 psig pressure
volume
250 μL
Related Categories
Legal Information
GASTIGHT is a registered trademark of Hamilton Co.
Hamilton is a registered trademark of Hamilton Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)