Skip to Content
Merck
All Photos(3)

Key Documents

H33207

Sigma-Aldrich

5-Hydroxyisoquinoline

technical grade, 90%

Synonym(s):

5-Isoquinolinol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H7NO
CAS Number:
Molecular Weight:
145.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

90%

SMILES string

Oc1cccc2cnccc12

InChI

1S/C9H7NO/c11-9-3-1-2-7-6-10-5-4-8(7)9/h1-6,11H

InChI key

CSNXUYRHPXGSJD-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Soomin Hyun et al.
Dalton transactions (Cambridge, England : 2003), 48(29), 10927-10932 (2019-06-04)
Self-assembly of ZnI2 with di(isoquinolin-5-yl)isophthalate (L) as a new bidentate N-donor gives rise to discrete crown-shaped M2L2 cyclodimers, 3.5CH3CN·0.7THF@[Zn2I4L2]. These metallacyclodimers are composed of a characteristically eclipsed molecular array, resulting in the formation of unusual 10.0 × 15.1 Å2 diameter
Keith L Constantine et al.
Journal of medicinal chemistry, 51(19), 6225-6229 (2008-09-06)
Fragment-like inhibitors of mitogen-activated protein kinase-activated protein kinase 2 (MK2) include 5-hydroxyisoquinoline (IC50 approximately 85 microM). Modeling studies identified four possible binding modes for this compound. Two-dimensional (1)H-(1)H NOESY data obtained with selectively protonated samples of MK2 in complex with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service