GF96939253
Stainless Steel - AISI 316L alloy, FeCr18Ni10Mo3
foil, 100 x 100mm, 0.5mm thickness, annealed, polished on both sides
Synonym(s):
FF210351
About This Item
Recommended Products
form
annealed (polished on both sides)
foil
manufacturer/tradename
Goodfellow 969-392-53
size × thickness
100 mm × 100 mm × 0.5 mm
Looking for similar products? Visit Product Comparison Guide
1 of 4
This Item | GF98629352 | GF10003239 | GF92991987 |
---|---|---|---|
annealed (polished on both sides), foil | annealed (polished on both sides), foil | annealed (polished on both sides), foil | annealed (polished on both sides), foil |
100 mm × 100 mm × 0.5 mm | 250 mm × 300 mm × 0.5 mm | 50 mm × 50 mm × 0.5 mm | 150 mm × 250 mm × 0.5 mm |
General description
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service