Skip to Content
Merck
All Photos(2)

Key Documents

GF02507116

Zinc

rod, 4.8 mm diameter, length 100 mm, high purity 99.99+%

Synonym(s):

ZN007905

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Zn
CAS Number:
Molecular Weight:
65.39
MDL number:
UNSPSC Code:
12141750
PubChem Substance ID:
NACRES:
NA.23

Pricing and availability is not currently available.

vapor pressure

1 mmHg ( 487 °C)

Assay

≥99.99%

form

rod

manufacturer/tradename

Goodfellow 025-071-16

resistivity

5.8 μΩ-cm, 20°C

L

100 mm

diameter

4.8 mm

bp

907 °C (lit.)

mp

420 °C (lit.)

density

7.133 g/mL at 25 °C (lit.)

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
GF09292079GF45701041GF21965609
Zinc rod, 4.8 mm diameter, length 100 mm, high purity 99.99+%

GF02507116

Zinc

Zinc rod, 4.8 mm diameter, length 1000 mm, high purity 99.99+%

GF09292079

Zinc

Zinc rod, 8 mm diameter, length 100 mm, high purity 99.999%

GF45701041

Zinc

Zinc rod, 8 mm diameter, length 50 mm, high purity 99.999%

GF21965609

Zinc

assay

≥99.99%

assay

≥99.99%

assay

99.999%

assay

99.999%

manufacturer/tradename

Goodfellow 025-071-16

manufacturer/tradename

Goodfellow 092-920-79

manufacturer/tradename

Goodfellow 457-010-41

manufacturer/tradename

Goodfellow 219-656-09

density

7.133 g/mL at 25 °C (lit.)

density

7.133 g/mL at 25 °C (lit.)

density

7.133 g/mL at 25 °C (lit.)

density

7.133 g/mL at 25 °C (lit.)

mp

420 °C (lit.)

mp

420 °C (lit.)

mp

420 °C (lit.)

mp

420 °C (lit.)

resistivity

5.8 μΩ-cm, 20°C

resistivity

5.8 μΩ-cm, 20°C

resistivity

5.8 μΩ-cm, 20°C

resistivity

5.8 μΩ-cm, 20°C

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


  • Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    Sorry, we don't have COAs for this product available online at this time.

    If you need assistance, please contact Customer Support.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service