BAF131643848
Scienceware® biohazard disposal bags
width × H × thickness 970 mm (38 in.) × 48 in. (1220 mm) × 0.04 mm (1.5 mil), pack of 100 ea, red
Synonym(s):
biohaz waste bag, biohazard disposal bags, waste bag
All Photos(2)
About This Item
UNSPSC Code:
41122800
NACRES:
NB.35
Recommended Products
material
red
packaging
pack of 100 ea
manufacturer/tradename
Bel-Art F13164-3848
capacity
151.4-189.3 L (40-50 gal)
width × H × thickness
970 mm (38 in.) × 48 in. (1220 mm) × 0.04 mm (1.5 mil)
Related Categories
General description
Bright red polypropylene bags include black biohazard symbol and precautionary procedures printed in four languages (English, Spanish, French, German) for added safety.
Features and Benefits
Bright red polypropylene bags include black biohazard symbol and precautionary procedures printed in four languages (English, Spanish, French, German) for added safety.
- Sterilization indicator patch darkens when exposed to steam sterilization for 20 minutes
- High temperature steam autoclavable to 135°C (275°F)
- Pass ASTM 1922 Tear Resistance and ASTM 1709 Dart Impact Tests
- Made in USA
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)