Skip to Content
Merck
All Photos(2)

Documents

A65158

Sigma-Aldrich

4-(Aminomethyl)piperidine

greener alternative

96%

Synonym(s):

4-AMP, 4-Piperidinemethanamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14N2
CAS Number:
Molecular Weight:
114.19
Beilstein:
471185
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

form

solid

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.49 (lit.)

bp

200 °C (lit.)

mp

25 °C (lit.)

greener alternative category

SMILES string

NCC1CCNCC1

InChI

1S/C6H14N2/c7-5-6-1-3-8-4-2-6/h6,8H,1-5,7H2

InChI key

LTEKQAPRXFBRNN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

4-(Aminomethyl)piperidine (4-AMP) can be used in the synthesis of Schiff bases. It is a trifunctional amine for the preparation of linear poly(amido amine)s, which form micelles for controlled delivery of drugs. 4-AMP can also be used as a linker for the synthesis of dendron-OMS hybrids.
Reactant:
  • Evaulated as a small molecule inhibitor of CD4-gp120 binding

Reagent
  • Involved in solvothermal preparation of germanium oxide hydroxide 3D net zeotype
  • Involved in zeolite synthesis

Coupling agent for adhesive systems

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

172.4 °F - closed cup

Flash Point(C)

78 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

p H-and Redox-R esponsive Poly (ethylene glycol) and Cholesterol-C onjugated Poly (amido amine) s Based Micelles for Controlled Drug Delivery
Cheng W, et al.
Macromolecular Bioscience, 14(3), 347-358 (2014)
Synthesis, characterization, antimicrobial activity, and optical properties of Schiff bases derived from 4-(Aminomethyl) Piperidine
Karthik CS, et al.
Indian Journal of Advances in Chemical Science, 206, 212-212 (2016)
Catalytic properties of dendron-OMS hybrids
Wang Q, et al.
J. Catal., 269(1), 15-25 (2010)
Lingling Mao et al.
Journal of the American Chemical Society, 140(10), 3775-3783 (2018-02-22)
The three-dimensional hybrid organic-inorganic perovskites have shown huge potential for use in solar cells and other optoelectronic devices. Although these materials are under intense investigation, derivative materials with lower dimensionality are emerging, offering higher tunability of physical properties and new

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service