Skip to Content
Merck
All Photos(1)

Key Documents

929883

Sigma-Aldrich

Platinum (Pt) Mesh Electrode

size 2.0 cm × 2.0 cm, 0.1 mm wire diameter, 99.9%

Synonym(s):

Platinum Mesh Electrode

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352103
NACRES:
NB.61
Pricing and availability is not currently available.

Assay

99.9%

size

2.0 cm × 2.0 cm

Application

This large area (2.0 cm x 2.0 cm , 0.1 mm wire diameter) and high purity (99.9%) platinum electrode has a surface area of mesh 500 sq. mm.

Legal Information

Product of Bioanalytical Systems, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. What is the thickness of the Pt electrode?

    1 answer
    1. The wires making up the 2cm x 2cm mesh have a diameter of 0.1mm. The adhesive that connects the mesh to the platinum wire has a variable thickness and may range from 2 - 4 mm.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service