Skip to Content
Merck
All Photos(1)

Key Documents

912824

Sigma-Aldrich

Butane-1,4-diammonium iodide

Synonym(s):

1,4-Butanediamine dihydriodide, 1,4-Butanediamine dihydroiodide, 1,4-Diaminobutane dihydriodide, Butane-1,4-diaminium iodide, Butane-1,4-diamino dihydriodide, Greatcell Solar®, NBUDAI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H14I2N2
CAS Number:
Molecular Weight:
343.98
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.23

form

powder

Quality Level

color

beige

SMILES string

I.I.NCCCCN

InChI key

XZUCBFLUEBDNSJ-UHFFFAOYSA-N

Application

Butane-1,4-diammonium iodide is a precursor for the synthesis of perovskite semiconductors in perovskite solar cells. Its addition to the precursor solution has been shown in increasing efficiency and stability of solar cells.
Organohalide based perovskites have emerged as an important class of material for solar cell applications. The variations/substitution in organohalide cations and anions is employed for the optimization of the band gap, carrier diffusion length, and power conversion efficiency of perovskites based solar cells. Therefore adding a suite of organohalide precursors would be a good expansion of our Perovskites portfolio.

Legal Information

Product of Greatcell Solar Materials Pty Ltd.
Greatcell Solar is a registered trademark of Greatcell Solar Materials Pty Ltd.
Greatcell Solar is a registered trademark of Greatcell Solar

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Limeng Ni et al.
ACS nano, 11(11), 10834-10843 (2017-10-25)
Self-assembled hybrid perovskite quantum wells have attracted attention due to their tunable emission properties, ease of fabrication, and device integration. However, the dynamics of excitons in these materials, especially how they couple to phonons, remains an open question. Here, we
Systematic studies on chain lengths, halide species, and well thicknesses for lead halide layered perovskite thin films.
Takeoka Y, et al.
Bulletin of the Chemical Society of Japan, 79(10), 1607-1613 (2006)
Qi Zhang et al.
Advanced materials (Deerfield Beach, Fla.), 30(18), e1704055-e1704055 (2018-03-27)
2D organic-inorganic hybrid perovskites (OIHPs) represent a unique class of materials with a natural quantum-well structure and quasi-2D electronic properties. Here, a versatile direct solution-based synthesis of mono- and few-layer OIHP nanosheets and a systematic study of their electronic structure

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service