Skip to Content
Merck
All Photos(2)

Key Documents

900441

Sigma-Aldrich

Green light-emitting spiro-copolymer

Synonym(s):

Livilux® SPG-01T

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

ribbon

Quality Level

mol wt

average Mn >100,000
average Mw >400,000

Application

Features:
  • Soluble green light-emitting spiro-copolymer.
  • High efficiency.
  • Long lifetime, with further potential to improve.
  • Saturated green color coordinates, with low operation voltage.

Typical performance measured in standard device stack (ITO/PEDOT:PSS 20 nm/SPG-01T 80 nm/Ba 6 nm/Ag 150 nm):
  • Max. efficiency: 11 Cd/A.
  • V_on: 2.5 V.
  • V at 100 cd/m2: 3.6 V.
  • V at 1000 cd/m2: 6.0 V.
  • Max. power eff.: 12 lm/W.
  • External quantum eff.: 4.1%.
  • CIE1931 at 100 cd/m2: (0.33, 0.59).
  • DC lifetime at 100 cd/m2 at RT: >10k hrs.
  • AC lifetime at 200 cd/m2 at RT: >5000 hrs.

Preparation Note

  • 10 g/L in toluene for spin-coating.
  • Special formulation needed for inkjet.
  • Preparation should be in N2 atmosphere.

Legal Information

Product of EMD Performance Materials Corp., an Affiliate of Merck KGaA, Darmstadt, Germany
Livilux is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Organic electronics utilizes organic conductors and semiconductors for applications in organic photovoltaics, organic light-emitting diodes, and organic field-effect transistors.

Organic electronics utilizes organic conductors and semiconductors for applications in organic photovoltaics, organic light-emitting diodes, and organic field-effect transistors.

Organic electronics utilizes organic conductors and semiconductors for applications in organic photovoltaics, organic light-emitting diodes, and organic field-effect transistors.

Organic electronics utilizes organic conductors and semiconductors for applications in organic photovoltaics, organic light-emitting diodes, and organic field-effect transistors.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service