Skip to Content
Merck
All Photos(1)

Key Documents

731749

Sigma-Aldrich

2,4,6-Triisopropylphenylmagnesium bromide solution

0.5 M in THF

Synonym(s):

Bromo[2,4,6-tris(1-methylethyl)phenyl]magnesium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H23BrMg
Molecular Weight:
307.55
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

solution

Quality Level

reaction suitability

reaction type: Grignard Reaction

concentration

0.5 M in THF

density

0.949 g/mL at 25 °C

SMILES string

CC(C)c1cc(C(C)C)c([Mg]Br)c(c1)C(C)C

InChI

1S/C15H23.BrH.Mg/c1-10(2)13-7-14(11(3)4)9-15(8-13)12(5)6;;/h7-8,10-12H,1-6H3;1H;/q;;+1/p-1

InChI key

LXUAJPDGIIDXFR-UHFFFAOYSA-M

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

-22.0 °F

Flash Point(C)

-30 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service