Skip to Content
Merck
All Photos(1)

Key Documents

714348

Sigma-Aldrich

Potassium oleate-13C18

endotoxin tested, 99 atom % 13C, 98% (CP)

Synonym(s):

Oleic acid-13C18 potassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13CH3(13CH2)713CH=13CH(13CH2)713CO2K
Molecular Weight:
338.42
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

isotopic purity

99 atom % 13C

Assay

98% (CP)

form

solid

suitability

endotoxin tested

mass shift

M+18

storage temp.

2-8°C

SMILES string

O=[13C]([O-])[13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13CH2]/[13CH]=[13CH]\[13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13CH3].[K+]

InChI

1S/C18H34O2.K/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20;/h9-10H,2-8,11-17H2,1H3,(H,19,20);/q;+1/p-1/b10-9-;/i1+1,2+1,3+1,4+1,5+1,6+1,7+1,8+1,9+1,10+1,11+1,12+1,13+1,14+1,15+1,16+1,17+1,18+1;

InChI key

MLICVSDCCDDWMD-DMVMXKRLSA-M

Looking for similar products? Visit Product Comparison Guide

Application

For R&D use only. Not for drug, household, or other uses. Any other use is the sole responsibility of the end user.

Endotoxin testing is done on the bulk form of the product before subdivision packaging. This bulk test does not guarantee that the subdivision or repackaged aliquot is endotoxin-free when it is received or used by the customer and does not imply suitability for any particular purpose. If the product must be endotoxin-free for the intended application, the product should be tested prior to actual use.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

284.0 °F - closed cup

Flash Point(C)

140 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service