Skip to Content
Merck
All Photos(1)

Documents

690309

Sigma-Aldrich

Sodium dicyanamide

Arxada quality, ≥97.0 % (w/w) (T)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaN(CN)2
CAS Number:
Molecular Weight:
89.03
Beilstein:
3913818
EC Number:
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.22

form

powder

Quality Level

quality

Arxada quality

manufacturer/tradename

Arxada AG

concentration

≥97.0 % (w/w) (T)

impurities

≤0.5 % (w/w) water (Karl Fischer)
≤3.0 % (w/w) NaCl (AT)

SMILES string

[Na]N(C#N)C#N

InChI

1S/C2N3.Na/c3-1-5-2-4;/q-1;+1

InChI key

IXBPPZBJIFNGJJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for preparation of:
  • Nickel diacetylpyridine dioximate pentanuclear, octanuclear and decanuclear cluster complexes
  • Decanuclear manganese(II/III) oxo azide diolate clusters as supertetrahedral building-blocks to design extended magnetic networks
  • Ruthenium bipyridine complexes entrapped in iron and manganese dicyanamide two-dimensional anionic sheet-like coordination polymeric networks exhibiting dual emission photoluminescence
  • Lead dicyanamide-bridged nitronyl nitroxide radical polymeric complex
  • Antiferromagnetically coupled hexanuclear copper(II) Schiff base complexes with dicyanamido bridges
  • CuII2L based polymeric ladders using dicyanamide bridges
  • Luminous hexacoordinated coordination polymers of cadmium(II), containing dicyanamide bridges

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel Rauber et al.
Physical chemistry chemical physics : PCCP, 19(40), 27251-27258 (2017-10-11)
Ionic liquids (ILs) exhibit tunable behaviour and properties that are due to their supramolecular structure. We synthesized a series of alkylated and fluorinated phosphonium dicyanamide ILs to study the relation between molecular structure and assembly with a focus on the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service