Skip to Content
Merck
All Photos(1)

Documents

689777

Sigma-Aldrich

O-[N-(3-Maleimidopropionyl)aminoethyl]-O′-[3-(N-succinimidyloxy)-3-oxopropyl]heptacosaethylene glycol

≥90% (oligomer purity)

Synonym(s):

MAL-PEG NHS, Maleimidopropionyl-PEG NHS ester (n=27)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C70H127N3O35
Molecular Weight:
1570.76
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥90% (oligomer purity)

form

powder

reaction suitability

reagent type: cross-linking reagent

impurities

≤2% hydrolyzed product

Ω-end

NHS ester

α-end

maleimide

storage temp.

−20°C

SMILES string

O=C(CCN1C(=O)C=CC1=O)NCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCC(=O)ON2C(=O)CCC2=O

InChI

1S/C70H127N3O35/c74-65(5-8-72-66(75)1-2-67(72)76)71-7-10-81-12-14-83-16-18-85-20-22-87-24-26-89-28-30-91-32-34-93-36-38-95-40-42-97-44-46-99-48-50-101-52-54-103-56-58-105-60-62-107-64-63-106-61-59-104-57-55-102-53-51-100-49-47-98-45-43-96-41-39-94-37-35-92-33-31-90-29-27-88-25-23-86-21-19-84-17-15-82-13-11-80-9-6-70(79)108-73-68(77)3-4-69(73)78/h1-2H,3-64H2,(H,71,74)

InChI key

QTIQGDBALCIANR-UHFFFAOYSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Progress in biotechnology fields such as tissue engineering and drug delivery is accompanied by an increasing demand for diverse functional biomaterials. One class of biomaterials that has been the subject of intense research interest is hydrogels, because they closely mimic the natural environment of cells, both chemically and physically and therefore can be used as support to grow cells. This article specifically discusses poly(ethylene glycol) (PEG) hydrogels, which are good for biological applications because they do not generally elicit an immune response. PEGs offer a readily available, easy to modify polymer for widespread use in hydrogel fabrication, including 2D and 3D scaffold for tissue culture. The degradable linkages also enable a variety of applications for release of therapeutic agents.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service