Skip to Content
Merck
All Photos(2)

Key Documents

684031

Sigma-Aldrich

(1S, 2S)-1,2-Bis(4-nitrophenyl)ethylenediamine dihydrochloride

97%

Synonym(s):

(1S, 2S)-1,2-Bis(4-nitrophenyl)-1,2-ethanediamine dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H14N4O4 · 2HCl
CAS Number:
Molecular Weight:
375.21
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

optical activity

[α]22/D -84.0°, c = 1 in H2O

mp

202-207 °C

SMILES string

Cl.Cl.N[C@H]([C@@H](N)c1ccc(cc1)[N+]([O-])=O)c2ccc(cc2)[N+]([O-])=O

InChI

1S/C14H14N4O4.2ClH/c15-13(9-1-5-11(6-2-9)17(19)20)14(16)10-3-7-12(8-4-10)18(21)22;;/h1-8,13-14H,15-16H2;2*1H/t13-,14-;;/m0../s1

InChI key

PDPYGNJVCKPVGM-AXEKQOJOSA-N

Application

(1S, 2S)-1,2-Bis(4-nitrophenyl)ethylenediamine dihydrochloride can be used to prepare a fluorous Fe-salen complex, which is used as a catalyst in the asymmetric epoxidation of C-C double bonds.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Unprecedented Asymmetric Epoxidation of Isolated Carbon-Carbon Double Bonds by a Chiral Fluorous Fe (III) Salen Complex: Exploiting Fluorophilic Effect for Catalyst Design
Kobayashi Y, et al.
European Journal of Organic Chemistry, 2019(13), 2401-2408 (2019)

Articles

Chiral vicinal diamines are of tremendous interest to the synthetic chemist as they are found in many chiral catalysts and pharmaceuticals.

Related Content

The Chin group is interested in computational and experimental approaches to understanding stereoselective recognition and catalysis. Their studies in weak forces (H-bonding, electronic and steric effects) has led to a highly efficient method for making limitless varieties of chiral vicinal diamines from the 'mother diamine' that are useful for developing stereoselective organocatalysts or transition metal-based catalysts as well as for developing drugs (Acc Chem Res (2012) p1345). The 'mother diamine' is also useful for making binol, monophos and binap analogs. The Chin group is also interested in using reversible covalent bonds for stereoselective recognition and L to D conversion of natural and non-natural amino acids (EJOC (2012) p229).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service