Skip to Content
Merck
All Photos(1)

Key Documents

677574

Sigma-Aldrich

Cholesterol-25,26,26,26,27,27,27-d7

99 atom % D, 98% (CP)

Synonym(s):

Cholest-5-en-25,26,26,26,27,27,27-d7-3-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27D7H39O
CAS Number:
Molecular Weight:
393.70
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

isotopic purity

99 atom % D

Assay

98% (CP)

form

solid

mol wt

393.63 (atom % calculation)

technique(s)

mass spectrometry (MS): suitable

mp

147-149 °C

mass shift

M+7

SMILES string

[H][C@@]12[C@]([C@](CC[C@H](O)C3)(C)C3=CC2)([H])CC[C@@]4(C)[C@@]1([H])CC[C@]4([H])[C@]([H])(C)CCCC([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H]

InChI

1S/C27H46O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h9,18-19,21-25,28H,6-8,10-17H2,1-5H3/t19-,21+,22+,23-,24+,25+,26+,27-/m1/s1/i1D3,2D3,18D

InChI key

HVYWMOMLDIMFJA-IFAPJKRJSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christophe A C Freyre et al.
Molecular cell, 76(5), 811-825 (2019-10-20)
Physical contact between organelles is vital to the function of eukaryotic cells. Lipid droplets (LDs) are dynamic organelles specialized in lipid storage that interact physically with mitochondria in several cell types. The mechanisms coupling these organelles are, however, poorly understood

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service