Skip to Content
Merck
All Photos(1)

Key Documents

64685

Sigma-Aldrich

Methoxyacetaldehyde dimethyl acetal

≥98.0% (GC)

Synonym(s):

1,1,2-Trimethoxyethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OCH2CH(OCH3)2
CAS Number:
Molecular Weight:
120.15
Beilstein:
741876
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (GC)

refractive index

n20/D 1.392 (lit.)
n20/D 1.393

bp

56-59 °C/56 mmHg (lit.)

density

0.932 g/mL at 25 °C (lit.)

SMILES string

COCC(OC)OC

InChI

1S/C5H12O3/c1-6-4-5(7-2)8-3/h5H,4H2,1-3H3

InChI key

DYOZNCVZPFIXLU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

72.5 °F - closed cup

Flash Point(C)

22.5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service