Skip to Content
Merck
All Photos(3)

Key Documents

642436

Sigma-Aldrich

2-Hydrazinobenzothiazole

97%

Synonym(s):

Benzothiazol-2-yl-hydrazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7N3S
CAS Number:
Molecular Weight:
165.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

198-202 °C (lit.)

functional group

hydrazine

SMILES string

NNc1nc2ccccc2s1

InChI

1S/C7H7N3S/c8-10-7-9-5-3-1-2-4-6(5)11-7/h1-4H,8H2,(H,9,10)

InChI key

JYSUYJCLUODSLN-UHFFFAOYSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lixiang Cheng et al.
BMC plant biology, 16(1), 188-188 (2016-09-01)
Drought stress is one of the most adverse environmental constraints to plant growth and productivity. Comparative proteomics of drought-tolerant and sensitive wheat genotypes is a strategy to understand the complexity of molecular mechanism of wheat in response to drought. This
Zhengyi Chen et al.
Analytica chimica acta, 1065, 29-39 (2019-04-22)
It remains a significant challenge for fast and high-throughput detection of trace analytes in complex samples with surface-enhanced Raman scattering (SERS) strategy due to the severe interference from matrices. In this work, a miniaturized array gas membrane separation (AGMS) device
M Beljean-Leymarie et al.
Analytical biochemistry, 173(1), 174-184 (1988-08-15)
A method for the measurement of malondialdehyde (MDA) using GLC with a nitrogen phosphorus detector (GLC/NPD) is described and evaluated. The method uses 2-hydrazinobenzothiazole (HBT) which forms condensation derivatives with MDA, acetoacetaldehyde, and acetylacetone (AA). GC/MS and 13C NMR studies

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service