Skip to Content
Merck
All Photos(1)

Documents

614696

Sigma-Aldrich

2,2,3,3,3-Pentafluoropropan-d2-ol

98 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3CF2CD2OH
Molecular Weight:
152.06
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

isotopic purity

98 atom % D

mass shift

M+2

SMILES string

[2H]C([2H])(O)C(F)(F)C(F)(F)F

InChI

1S/C3H3F5O/c4-2(5,1-9)3(6,7)8/h9H,1H2/i1D2

InChI key

PSQZJKGXDGNDFP-DICFDUPASA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service