Skip to Content
Merck
All Photos(1)

Documents

575798

Sigma-Aldrich

Poly(pyromellitic dianhydride-co-4,4′-oxydianiline), amic acid solution

12.8 wt. % (80% NMP/20% aromatic hydrocarbon)

Synonym(s):

Pyre-M.L.® RC-5069, Polyamic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C12H12N2O .C10H2O6)x
UNSPSC Code:
12162002
NACRES:
NA.23

Quality Level

concentration

12.8 wt. % (80% NMP/20% aromatic hydrocarbon)

refractive index

n20/D 1.5050

viscosity

5-12 poise

solubility

H2O: soluble 65-71 wt. %

density

1.085 g/mL at 25 °C

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

  • Paper Title: Synthesis of nano-polyimide for microelectronic applications. This study discusses the synthesis processes from Poly (amic acid) (PAA) of poly (pyromellitic dianhydride-co-4,4′-oxydianiline) and its applications in microelectronics, highlighting its potential in advanced fabrication techniques (Ebnalwaled et al., 2016).

Legal Information

Pyre-M.L. is a registered trademark of Industrial Summit Technologies Corp.

Signal Word

Danger

Hazard Classifications

Asp. Tox. 1 - Carc. 1B - Eye Irrit. 2 - Flam. Liq. 3 - Muta. 1B - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

138.2 °F - closed cup

Flash Point(C)

59 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service