Skip to Content
Merck
All Photos(1)

Key Documents

560510

Sigma-Aldrich

3-(Diethylamino)propyl isothiocyanate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2)2N(CH2)3NCS
CAS Number:
Molecular Weight:
172.29
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.5000 (lit.)

density

0.954 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCN(CC)CCCN=C=S

InChI

1S/C8H16N2S/c1-3-10(4-2)7-5-6-9-8-11/h3-7H2,1-2H3

InChI key

QPBVKSLJBRCKIF-UHFFFAOYSA-N

Gene Information

Related Categories

Application

3-(Diethylamino)propyl isothiocyanate (DEAP) may be used to synthesize GCS-g-DEAP [glycol chitosan (GCS) grafted with DEAP groups].

Caution

Refrigerate

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nam Muk Oh et al.
Colloids and surfaces. B, Biointerfaces, 78(1), 120-126 (2010-03-20)
In this study, a novel pH-responsive nanogel composed of glycol chitosan (GCS) grafted with functional 3-diethylaminopropyl (DEAP) groups (denoted as GCS-g-DEAP hereafter) was fabricated. The GCS-g-DEAP was designed to have a self-assembled arrangement consisting of hydrophilic block (GCS) and hydrophobic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service