Skip to Content
Merck
All Photos(1)

Documents

534749

Sigma-Aldrich

2-(4-Bromophenyl)oxirane

96%

Synonym(s):

4-Bromostyrene oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7BrO
CAS Number:
Molecular Weight:
199.04
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

bp

91-93 °C/2 mmHg (lit.)

mp

26-29 °C (lit.)

SMILES string

Brc1ccc(cc1)C2CO2

InChI

1S/C8H7BrO/c9-7-3-1-6(2-4-7)8-5-10-8/h1-4,8H,5H2

InChI key

NNINSLOEPXEZOZ-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

102.0 °F - closed cup

Flash Point(C)

38.89 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service