437948
2-Chloro-1,1,1-trimethoxyethane
97%
Synonym(s):
Trimethyl chloro-orthoacetate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
ClCH2C(OCH3)3
CAS Number:
Molecular Weight:
154.59
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
refractive index
n20/D 1.425 (lit.)
bp
138 °C (lit.)
density
1.147 g/mL at 25 °C (lit.)
functional group
chloro
ether
SMILES string
COC(CCl)(OC)OC
InChI
1S/C5H11ClO3/c1-7-5(4-6,8-2)9-3/h4H2,1-3H3
InChI key
NPEIUNVTLXEOLT-UHFFFAOYSA-N
Related Categories
General description
2-Chloro-1,1,1-trimethoxyethane is a chloromethyl heterocyclic compound. It participates in the synthesis of 2-phenyl-1,3,4-oxadiazole derivatives.
Application
2-Chloro-1,1,1-trimethoxyethane may be used for the synthesis of chlorotriazolinone, an NK1 antagonist and 2-chloromethyl-7-[3-(trifluoromethyl)pyridin-2-yl]-3H-quinazolin-4-one.
Signal Word
Warning
Hazard Statements
Hazard Classifications
Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
109.4 °F - closed cup
Flash Point(C)
43 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Elfatih Elzein et al.
Bioorganic & medicinal chemistry letters, 14(24), 6017-6021 (2004-11-18)
New inhibitors of palmitoyl-CoA oxidation are based on the introduction of nitrogen heterocycles in the 'Western Portion' of the molecule. SAR studies led to the discovery of CVT-4325 (shown), a potent FOXi (IC50=380 nM rat mitochondria) with favorable PK properties
Daisy van Veghel et al.
ACS chemical neuroscience, 4(4), 624-634 (2013-02-21)
The transient receptor potential vanilloid subfamily member 1 (TRPV1) cation channel is known to be involved in pain nociception and neurogenic inflammation, and accumulating evidence suggests that it plays an important role in several central nervous system (CNS)-related disorders. TRPV1-specific
A new synthesis of 1, 2, 4-triazolin-5-ones: application to the convergent synthesis of an NK 1 antagonist.
Cowden CJ, et al.
Tetrahedron Letters, 41(44), 8661-8664 (2000)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service