Skip to Content
Merck
All Photos(3)

Key Documents

416002

Sigma-Aldrich

Poly(acrylic acid) solution

average Mw ~250,000, 35 wt. % in H2O

Synonym(s):

PAA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C3H4O2)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

viscous liquid

mol wt

average Mw ~250,000

concentration

35 wt. % in H2O

refractive index

n20/D 1.395

density

1.150 g/mL at 25 °C

SMILES string

OC(=O)C=C

InChI

1S/C3H4O2.Na/c1-2-3(4)5;/h2H,1H2,(H,4,5);/q;+1/p-1

InChI key

NNMHYFLPFNGQFZ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Poly(acrylic acid) solution (PAA) is an anionic polymer that can be synthesized by the free radical polymerization of acrylic acid. It has a swelling nature that tends to absorb and retain the water. Its high ion exchange capacity makes it useful in the formation of membranes.

Application

PAA can be used for a variety of applications such as gas sensors, electrical devices, and cosmetics.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrospun fibrous polyacrylic acid membrane-based gas sensors
Ding B, et al.
Sensors and Actuators B, Chemical, 106(1), 477-483 (2005)
Poly (acrylic acid) nanofibers by electrospinning
Kim B, et al.
Materials Letters, 59(7), 829-832 (2005)
Water absorption, retention and the swelling characteristics of cassava starch grafted with polyacrylic acid
Witono JR, et al.
Carbohydrate Polymers, 103(1), 325-332 (2014)
Swelling behavior of acrylic acid hydrogels prepared by gamma-radiation crosslinking of polyacrylic acid in aqueous solution
Jabbari E and Nozari S
European Polymer Journal, 36(12), 2685-2692 (2000)
Fast adsorption of methylene blue on polyacrylic acid-bound iron oxide magnetic nanoparticles
Mak S and Chen D
Dyes and Pigments, 61(1), 93-98 (2004)

Articles

Laboratory safety tools: Offers educational tools for chemistry, prioritizing customer health and safety, with online support available.

Laboratory safety tools: Offers educational tools for chemistry, prioritizing customer health and safety, with online support available.

Laboratory safety tools: Offers educational tools for chemistry, prioritizing customer health and safety, with online support available.

We have discussed shortly the applications of 1:1 and 2:1 phyllosilicate material in membrane technology, as well as the basic geometric understanding of their layered structures.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service